missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB11FIP4A Polyclonal Antibody
GREENER_CHOICE

Product Code. 17117618 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17117618 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17117618 Supplier Invitrogen™ Supplier No. PA5116379

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAB11FIP4A Polyclonal Antibody for Western Blot

Acts as a regulator of endocytic traffic by participating in membrane delivery. Required for the abcission step in cytokinesis, possibly by acting as an 'address tag' delivering recycling endosome membranes to the cleavage furrow during late cytokinesis. May play a role in differentiation during retinal development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB11FIP4A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene rab11fip4a
Gene Accession No. Q3LGD4
Gene Alias FIP4-Rab11; hypothetical protein LOC436806; RAB11 family interacting protein 4 (class II) a; rab11 family-interacting protein 4A; Rab11-family interacting protein 4A; Rab11-family interacting protein 4B; rab11fip4; rab11fip4a; rab11-FIP4-A; zgc:92911; zRab11-FIP4-A
Gene Symbols rab11fip4a
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 183-216 amino acids from the Central region of DANRE rab11fip4a.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 436806
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.