missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15662-1-AP-20UL
This item is not returnable.
View return policy
Description
Localized to biosynthetic and endosomal compartments, Rab14 is involved in the biosynthetic/recycling pathway between the Golgi and endosomal compartments.Specifications
| RAB14 | |
| Polyclonal | |
| Unconjugated | |
| RAB14 | |
| FBP, RAB 14, RAB14, Ras related protein Rab 14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51552, 68365, 94197 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P61106, P61107, Q91V41 | |
| RAB14 | |
| RAB14 Fusion Protein Ag8092 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction