missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB17 Monoclonal Antibody (OTI3A5)
GREENER_CHOICE

Product Code. 15934724
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15934724 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15934724 Supplier Invitrogen™ Supplier No. MA525118

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RAB17 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The Rab subfamily of small GTPases plays an important role in the regulation of membrane trafficking. RAB17 is an epithelial cell-specific GTPase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB17
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3A5
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Rab17
Gene Accession No. Q9H0T7
Gene Alias AW413472; Rab17; RAB17, member RAS oncogene family; ras-related protein Rab-17; RGD1562478
Gene Symbols Rab17
Host Species Mouse
Immunogen Full length human recombinant protein of RAB17 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64284
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.