missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab1A Antibody (4G10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11043AF488
This item is not returnable.
View return policy
Description
Rab1A Monoclonal antibody specifically detects Rab1A in Human, Mouse samples. It is validated for Western Blot
Specifications
| Rab1A | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 4G10 | |
| Western Blot | |
| DKFZp564B163, member RAS oncogene family, RAB1A, member RAS oncogene family, ras-related protein Rab-1A | |
| Full length recombinant Rab1A protein (NP_004152.1) | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 5861 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction