missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab1A Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94057-0.1ml
This item is not returnable.
View return policy
Description
Rab1A Polyclonal antibody specifically detects Rab1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Rab1A | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| DKFZp564B163, member RAS oncogene family, RAB1A, member RAS oncogene family, ras-related protein Rab-1A | |
| A synthetic peptide corresponding to a sequence within amino acids 1-205 of human Rab1A (NP_004152.1). MSSMNPEYDYLFKLLLIGDSGVGKSCLLLRFADDTYTESYISTIGVDFKIRTIELDGKTIKLQIWDTAGQERFRTITSSYYRGAHGIIVVYDVTDQESFNNVKQWLQEIDRYASENVNKLLVGNKCDLTTKKVVDYTTAKEFADSLGIPFLETSAKNATNVEQSFMTMAAEIKKRMGPGATAGGAEKSNVKIQSTPVKQSGGGCC | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 5861 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction