missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46830
This item is not returnable.
View return policy
Description
Rab1A Polyclonal antibody specifically detects Rab1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Rab1A | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| P62820 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| DKFZp564B163, member RAS oncogene family, RAB1A, member RAS oncogene family, ras-related protein Rab-1A | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SAKNATNVEQSFMTMAAEIKKRMGPGA | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 5861 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction