Learn More
Invitrogen™ RAB1A Monoclonal Antibody (4G10), FITC
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545967
Product Code. 17827232
  • € 478.00 / 100µg
Description
A 1:1,000 dilution of MA5-45967 was sufficient for detection of RAB1A in 10 μg (A549, MEF); 20 μg (Mouse Brain) lysates by ECL immunoblot analysis. Detects approximately 23 kDa. Cross-reacts with RAB1B and RAB1C.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a member of the L18AE family of ribosomal proteins that is a component of the 60S subunit. The encoded protein may play a role in viral replication by interacting with the hepatitis C virus internal ribosome entry site (IRES). This gene is co-transcribed with the U68 snoRNA, located within the third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAB1A
Monoclonal
1 mg/mL
PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4
P62820, P62821
RAB1A
Full length recombinant RAB1A protein.
100 μg
Primary
Human, Mouse
Antibody
IgG1
Immunoprecipitation, Western Blot
4G10
FITC
RAB1A
Ac2-048; DKFZP564B163; Gtbp; GTP binding protein Rab1a; LOW QUALITY PROTEIN: ras-related protein Rab-1A; mKIAA3012; Rab GTPase YPT1 homolog; RAB1; Rab-1; RAB1, member RAS oncogene family; Rab1A; RAB1A, member RAS oncogene family; RAB1A-like protein; Rab1r; Ras-related protein Rab-1A; ras-related YPT1 protein; YPT1; YPT1-related protein
Mouse
Protein G
RUO
19324, 5861
4°C, store in dark