missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB1A Monoclonal Antibody (7H4), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545963
This item is not returnable.
View return policy
Description
A 1:1,000 dilution of MA5-45963 was sufficient for detection of RAB1A in 10 μg (A549, MEF); 20 μg (Mouse Brain) lysates by ECL immunoblot analysis. Detects approximately 23 kDa. Cross-reacts with RAB1B.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a member of the L18AE family of ribosomal proteins that is a component of the 60S subunit. The encoded protein may play a role in viral replication by interacting with the hepatitis C virus internal ribosome entry site (IRES). This gene is co-transcribed with the U68 snoRNA, located within the third intron. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed throughout the genome.
Specifications
| RAB1A | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| P62820, P62821 | |
| RAB1A | |
| Full length recombinant RAB1A protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| 7H4 | |
| APC | |
| RAB1A | |
| Ac2-048; DKFZP564B163; Gtbp; GTP binding protein Rab1a; LOW QUALITY PROTEIN: ras-related protein Rab-1A; mKIAA3012; Rab GTPase YPT1 homolog; RAB1; Rab-1; RAB1, member RAS oncogene family; Rab1A; RAB1A, member RAS oncogene family; RAB1A-like protein; Rab1r; Ras-related protein Rab-1A; ras-related YPT1 protein; YPT1; YPT1-related protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 19324, 5861 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction