missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB1B Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18251UV
This item is not returnable.
View return policy
Description
RAB1B Polyclonal antibody specifically detects RAB1B in Human, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| RAB1B | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| RAB1B, member RAS oncogene family, ras-related protein Rab-1B, small GTP-binding protein | |
| Synthetic peptide from the C-terminal of Human RAB1B (aa. 179-189) | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 81876 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction