missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab20 Antibody (OTI1F1), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73768AF700
This item is not returnable.
View return policy
Description
Rab20 Monoclonal specifically detects Rab20 in Human samples. It is validated for Western Blot.
Specifications
| Rab20 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RAB20 | |
| Full length recombinant protein of human Rab20 (NP_060287) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI1F1 | |
| Western Blot | |
| FLJ20429, RAB20, member RAS oncogene family, ras-related protein Rab-20 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 55647.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction