missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB21 Antibody (OTI6E12), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73769AF532
This item is not returnable.
View return policy
Description
RAB21 Monoclonal specifically detects RAB21 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| RAB21 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RAB21 | |
| Full length recombinant protein of human RAB21 (NP_055814) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6E12 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| KIAA0118ras-related protein Rab-21, RAB21, member RAS oncogene family | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 23011.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction