Learn More
ILK Monoclonal Antibody (OTI2H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804883
Encompass_Preferred
 View more versions of this product
Product Code. 17594633
  • € 540.87 / 100µL
Description
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
ILK
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13418
ILK
Human recombit protein fragment corresponding to amino acids 1-244 of human ILK produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Western Blot
OTI2H3
Unconjugated
ILK
HEL-S-28, ILK-1, ILK-2, P59, p59ILK
Mouse
Affinity Chromatography
RUO
3611
-20° C, Avoid Freeze/Thaw Cycles