Learn More
RAB21 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI6F6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505738
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RAB21 is a member of a subfamily of small GTP-binding protein genes of the Ras superfamily that plays an important role in intracellular vesicular targeting. In non-polarized Caco-2 cells, RAB21 showed a general endoplasmic reticulum (ER)-like localization, while in polarized cells, RAB21 localized to apical vesicles. RAB21 has been shown to associate with integrin subunits and to be important for receptor entry into cells via RAB5/RAB21 endosomes. The GTPase-activating protein p120RasGAP regulates cell motility by controlling the return of the endocytosed integrins to the plasma membrane by competing with RAB21 for binding to overlapping sites on the Alpha-tail of endocytosed integrin.Specifications
| RAB21 | |
| Monoclonal | |
| Unconjugated | |
| RAB21 | |
| KIAA0118 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100683312, 216344, 23011, 299799 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI6F6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P35282, P55745, Q6AXT5, Q9UL25 | |
| RAB21 | |
| Full length human recombit protein of human RAB21 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.