missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB22A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564298
This item is not returnable.
View return policy
Description
Immunogen sequence: ELFIEISRRI PSTDANLPSG GKGFKLRRQP SEPKRSCC Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 89%.
The protein encoded by this gene is a member of the RAB family of small GTPases. The GTP-bound form of the encoded protein has been shown to interact with early-endosomal antigen 1.
Specifications
| RAB22A | |
| Polyclonal | |
| Unconjugated | |
| RAB22A | |
| 3732413A17Rik; AI662177; AW319644; AW550514; E130120E14Rik; GTP-binding protein RAB22A; im:7136651; MGC16770; rab-14; Rab22; Rab-22; RAB22, member RAS oncogene family; Rab22a; Rab22a protein; RAB22A, member RAS oncogene family; RAS-related protein RAB-22A; zgc:101631 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57403 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UL26 | |
| RAB22A | |
| Recombinant protein corresponding to Human RAB22A. Recombinant protein control fragment (Product #RP-103645). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction