missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB23 Antibody (OTI4C7), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
€ 256.00 - € 550.00
Specifications
| Antigen | RAB23 |
|---|---|
| Clone | OTI4C7 |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:500 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306576
|
Novus Biologicals
NBP3-44949-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306413
|
Novus Biologicals
NBP3-44949-30ul |
30 μL |
€ 256.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAB23 Monoclonal antibody specifically detects RAB23 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| RAB23 | |
| Western Blot 1:2000, Immunohistochemistry 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp781H0695, HSPC137, MGC8900, RAB family small GTP binding protein RAB 23, RAB23, member RAS oncogene family, ras-related protein Rab-23 | |
| Human recombinant protein fragment corresponding to amino acids 1-234 of human RAB23(NP_899050) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI4C7 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Signal Transduction | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 51715 | |
| IgG1 | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title