missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB23 Monoclonal Antibody (OTI2A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809380S
This item is not returnable.
View return policy
Description
RAB23 Monoclonal antibody specifically detects RAB23 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| RAB23 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y6G3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 28977 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A8 | |
| Unconjugated | |
| MRPL42 | |
| AW545388; DKFZp781H0695; HSPC137; MGC8900; opb; opb2; open brain 2; OTTHUMP00000040021; protein open brain; RAB 23; RAB family small GTP binding protein RAB 23; rab-15; RAB23; RAB23, member RAS oncogene family; ras-related protein Rab-23; RP3-496N17.3 | |
| Full length human recombinant protein of human MRPL42 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction