missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab24 Antibody (OTI3H1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73770IR
This item is not returnable.
View return policy
Description
Rab24 Monoclonal specifically detects Rab24 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Rab24 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RAB24 | |
| Full length recombinant protein of human Rab24 (NP_001026847) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI3H1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| RAB24, member RAS oncogene family, ras-related protein Rab-24 | |
| Mouse | |
| Affinity Purified | |
| Autophagy, Signal Transduction | |
| 53917.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction