missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAB24 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16610525 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16610525 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16610525 Supplier Invitrogen™ Supplier No. PA5104070

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAB24.

RAB24 may be involved in autophagy-related processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAB24
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAB24
Gene Accession No. P35290, Q969Q5
Gene Alias 6530406O07Rik; rab-16; RAB24; RAB24, member RAS oncogene family; ras-related protein Rab-24
Gene Symbols RAB24
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAB24(Accession Q969Q5), corresponding to amino acid residues E126-A176.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19336, 361208, 53917
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.