missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB25 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149227
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
RAB proteins, such as RAB25, are members of the RAS superfamily of small GTPases that are involved in membrane trafficking. Members of the RAB11 subfamily, including RAB25, control the return of internalized membrane-associated moieties to the cell surface.
Specifications
| RAB25 | |
| Polyclonal | |
| Unconjugated | |
| RAB25 | |
| AV012978; CATX8; CATX-8; RAB11C; RAB25; RAB25, member RAS oncogene family; ras-related protein Rab-25 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 57111 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| P57735 | |
| RAB25 | |
| E. coli-derived recombinant human RAB25, Met1-Ala195. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction