Learn More
RAB27A Mouse anti-Human, CoraLite 594, Clone: 1C4B8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-66058-100UL
Product Code. 16870946
  • € 450.00 / 100µL
Description
The RAB27A antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human RAB27A. This antibody recognizes human antigen. The RAB27A antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
RAB27A
Monoclonal
CoraLite 594
100 μL
5873
IgG1
Immunofluorescence
1C4B8