missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB27B Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97109R
This item is not returnable.
View return policy
Description
RAB27B Polyclonal specifically detects RAB27B in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RAB27B | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| C25KG, RAB27B, member RAS oncogene family, ras-related protein Rab-27B | |
| Produced in rabbits immunized with purified, recombinant RAB27B (NP_004154.2; Thr2-Lys215). | |
| 0.1 mL | |
| Signal Transduction | |
| 5874 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction