missing translation for 'onlineSavingsMsg'
Få mere at vide

RAB27B Antibody [FITC], Novus Biologicals Biologicals™

Artikelnummer. 30488685 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mL
Pakningsstørrelse:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30488685 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30488685 Leverandør Novus Biologicals Leverandørnr. NBP335136F

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

RAB27B Polyclonal antibody specifically detects RAB27B in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen RAB27B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias C25KG, RAB27B, member RAS oncogene family, ras-related protein Rab-27B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-218 of human RAB27B (NP_004154.2).,, Sequence:, MTDGDYDYLIKLLALGDSGVGKTTFLYRYTDNKFNPKFITTVGIDFREKRVVYNAQGPNGSSGKAFKVHLQLWDTAGQERFRSLTTAFFRDAMGFLLMFDLTSQQSFLNVRNWMSQLQANAYCENPDIVLIGNKADLPDQREVNERQARELADKYGIPYFETSAATGQNVEKAVETLLDLIMKRMEQCVEKTQIPDTVNGGNSGNLDGEKPPEKKCIC
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5874
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.