Learn More
RAB27B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89538
This item is not returnable.
View return policy
Description
RAB27B Polyclonal specifically detects RAB27B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| C25KG, RAB27B, member RAS oncogene family, ras-related protein Rab-27B | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| RAB27B | |
| IgG |