missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB27B Monoclonal Antibody (1596CT584.1.45)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524743
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
RAB27B Monoclonal Antibody for Western Blot
RAB27B may be involved in targeting uroplakins to urothelial apical membranes.
Tekniske data
| RAB27B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O00194, Q99P58 | |
| RAB27B | |
| human RAB27B recombinant protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 1596CT584.1.45 | |
| Unconjugated | |
| RAB27B | |
| 2310021G14Rik; B130064M09Rik; C25KG; rab27b; RAB27B, member RAS oncogene family; ras-related protein Rab-27B; si:ch1073-112i17.1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5874, 80718 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion