missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB29 Polyclonal Antibody

Brand: Thermo Scientific PA5115819
This item is not returnable.
View return policy
Description
Specifications
| RAB29 | |
| Polyclonal | |
| Unconjugated | |
| RAB29 | |
| RAB29; RAB29, member RAS oncogene family; RAB7, member RAS oncogene family-like 1; RAB7L; RAB7L1; Rab-7-like protein 1; Ras-related GTP-binding protein Rab29; ras-related protein Rab-29; Ras-related protein Rab-7L1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 171122, 226422, 8934 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O14966, Q63481, Q91YQ1 | |
| RAB29 | |
| A synthesized peptide derived from human RAB29(Accession O14966), corresponding to amino acid residues W51-R101. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction