missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101823
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB2A.
Members of the Rab protein family are nontransforming monomeric GTP-binding proteins of the Ras superfamily that contain 4 highly conserved regions involved in GTP binding and hydrolysis. Rabs are prenylated, membrane-bound proteins involved in vesicular fusion and trafficking. The mammalian RAB proteins show striking similarities to the S. cerevisiae YPT1 and SEC4 proteins, Ras-related GTP-binding proteins involved in the regulation of secretion.
Specifications
| RAB2A | |
| Polyclonal | |
| Unconjugated | |
| Rab2a | |
| 9330148M11Rik; C80220; GTP binding protein; GTP-binding protein; LHX; OTTMUSP00000004796; rab 2; RAB2; RAB2, member RAS oncogene family; Rab2a; RAB2A, member RAS oncogene family; ras-related protein Rab-2A; RP23-183L6.1; small GTP binding protein RAB2A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5862, 59021, 65158 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05712, P53994, P61019 | |
| Rab2a | |
| A synthesized peptide derived from human RAB2A(Accession P61019), corresponding to amino acid residues L83-E133. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction