missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB30 Antibody (OTI3E7), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73771AF405
This item is not returnable.
View return policy
Description
RAB30 Monoclonal specifically detects RAB30 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| RAB30 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RAB30 | |
| Full length recombinant protein of human RAB30 (NP_055303) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3E7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| RAB30, member RAS oncogene family, ras-related protein Rab-30 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 27314.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction