missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB30 Monoclonal Antibody (OTI4B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505343
This item is not returnable.
View return policy
Description
RAB30 (Ras-related protein Rab-30) encodes for small GTPases Rab which are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. Required for maintaining the structural integrity of the Golgi apparatus, possibly by mediating interactions with cytoplasmic scaffolding proteins.Specifications
| RAB30 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15771 | |
| RAB30 | |
| Full length human recombit protein of human RAB30 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B9 | |
| Unconjugated | |
| RAB30 | |
| RAB30, member RAS oncogene family | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27314 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction