missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584462
This item is not returnable.
View return policy
Description
Immunogen sequence: SDNVEKLFLD LACRLISEAR QNTLVNNVSS PLPGEGKSIS YLTCCNF Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
RAB30 (Ras-related protein Rab-30) encodes for small GTPases Rab which are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. Required for maintaining the structural integrity of the Golgi apparatus, possibly by mediating interactions with cytoplasmic scaffolding proteins.
Specifications
| RAB30 | |
| Polyclonal | |
| Unconjugated | |
| RAB30 | |
| 5033421K01Rik; AI323892; EGK_06727; Rab30; RAB30, member RAS oncogene family; ras-related protein Rab-30; Rsb30 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27314 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15771 | |
| RAB30 | |
| Recombinant protein corresponding to Human RAB30. Recombinant protein control fragment (Product #RP-107019). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu