missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB31 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97073IR
This item is not returnable.
View return policy
Description
RAB31 Polyclonal specifically detects RAB31 in Human samples. It is validated for ELISA.
Specifications
| RAB31 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| DyLight 755 | |
| Rab22B, RAB31, member RAS oncogene family, Ras-related protein Rab-22B, ras-related protein Rab-31 | |
| Produced in rabbits immunized with purified, recombinant RAB31 (NP_006859.2; Met1-Cys195). | |
| 0.1 mL | |
| Signal Transduction | |
| 11031 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction