missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]ethanone Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
1-(4-Amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]ethanone Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 1-(4-Amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]ethanone Hydrochloride |
| CAS | 37148-49-5 |
| Analyte or Component Names | 1-(4-Amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]ethanone Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H16 Cl2 N2 O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanone, 1-(4-amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (9CI), Clenbuterol Hydrochloride Imp B (EP), Clenbuterol Imp B (EP), Clenbuterol USP Related Compound B, Clenbuterol USP RC B, Clenbuterol Related Compound B as Hydrochloride, Clenbuterol Hydrochloride Impurity B as Hydrochloride, Clenbuterol Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C12H16Cl2N2O.ClH/c1-12(2,3)16-6-10(17)7-4-8(13)11(15)9(14)5-7;/h4-5,16H,6,15H2,1-3H3;1H |
| SMILES | Cl.CC(C)(C)NCC(=O)c1cc(Cl)c(N)c(Cl)c1 |
| IUPAC Name | 1-(4-amino-3,5-dichlorophenyl)-2-(tert-butylamino)ethanone;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction