missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB33B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104074
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB33B.
Small GTP-binding proteins of the RAB family, such as RAB33B, play important roles at defined steps of vesicular transport in protein secretion and the endocytosis pathway (Zheng et al., 1998 ).
Specifications
| RAB33B | |
| Polyclonal | |
| Unconjugated | |
| RAB33B | |
| RAB33B; RAB33B, member of RAS oncogene family; RAB33B, member RAS oncogene family; ras-related protein Rab-33B; ras-related protein Rab-33B; LOW QUALITY PROTEIN: ras-related protein Rab-33B; SMC2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19338, 365793, 83452 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35963, Q9H082 | |
| RAB33B | |
| A synthesized peptide derived from human RAB33B(Accession Q9H082), corresponding to amino acid residues I129-A179. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction