missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103244
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB35.
RAB35 possesses GTPase activity.
Specifications
| RAB35 | |
| Polyclonal | |
| Unconjugated | |
| RAB35 | |
| 9530019H02Rik; AU040256; GTP-binding protein RAY; H-ray; I79_001225; RAB1C; Rab35; RAB35, member RAS oncogene family; Ras-related protein Rab-1C; ras-related protein rab-1c (GTP-binding protein ray); Ras-related protein Rab-35; RAY | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11021, 288700, 77407 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q15286, Q5U316, Q6PHN9 | |
| RAB35 | |
| A synthesized peptide derived from human RAB35(Accession Q15286), corresponding to amino acid residues N98-E148. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction