missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total RAB36.
Protein transport. Probably involved in vesicular traffic. [UniProt].
Specifications
Specifications
| Antigen | RAB36 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | RAB36 |
| Gene Accession No. | O95755, Q8CAM5 |
| Gene Alias | 6330593L16Rik; AW047545; Rab36; RAB36, member RAS oncogene family; ras-related protein Rab-36; small GTP-binding protein Rab36 |
| Gene Symbols | RAB36 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
