Learn More
RAB37 Antibody (OTI2F1), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-73772V
Product Code. 18120967
  • € 751.00 / 0.10mL
Description
RAB37 Monoclonal specifically detects RAB37 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAB37
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
RAB37
Full length recombinant protein of human RAB37 (NP_783865) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG2a
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI2F1
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
FLJ32507, member of RAS oncogene family, RAB37, member RAS oncogene family, ras-related protein Rab-37
Mouse
Affinity Purified
Signal Transduction
326624.0
Store at 4°C in the dark.