Learn More
RAB37 Monoclonal Antibody (OTI3B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505309S
Encompass_Preferred
 View more versions of this product
Product Code. 17513602
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
RAB37 Monoclonal antibody specifically detects RAB37 in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
RAB37
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96SF2
Mouse
Affinity chromatography
RUO
150160
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Immunocytochemistry, Western Blot
OTI3B2
Unconjugated
CCT8L2
B230331O03Rik; B230354I04Rik; GTPase Rab37; LOC100364984; Rab37; RAB37, member of RAS oncogene family; RAB37, member RAS oncogene family; ras-related protein Rab-37; rCG35079-like
Full length human recombinant protein of human CCT8L2 produced in HEK293T cell.
30 μL