Learn More
RAB37 Mouse anti-Human, Clone: OTI1E10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505339
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Rab proteins are low molecular mass GTPases that are critical regulators of vesicle trafficking. For additional background information on Rab proteins.Specifications
| RAB37 | |
| Monoclonal | |
| Unconjugated | |
| RAB37 | |
| RAB37, member RAS oncogene family | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 326624 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1E10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96AX2 | |
| RAB37 | |
| Full length human recombit protein of human RAB37 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.