missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544416
This item is not returnable.
View return policy
Description
Peptide sequence: ERVIRSEDGE TLAREYGVPF LETSAKTGMN VELAFLAIAK ELKYRAGHQA Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
Rab proteins are low molecular mass GTPases that are critical regulators of vesicle trafficking. For additional background information on Rab proteins.
Specifications
| RAB37 | |
| Polyclonal | |
| Unconjugated | |
| RAB37 | |
| B230331O03Rik; B230354I04Rik; GTPase Rab37; LOC100364984; Rab37; RAB37, member of RAS oncogene family; RAB37, member RAS oncogene family; ras-related protein Rab-37; rCG35079-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 326624 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96AX2 | |
| RAB37 | |
| Synthetic peptide directed towards the middle region of human RAB37. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction