missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101827
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB37.
Rab proteins are low molecular mass GTPases that are critical regulators of vesicle trafficking. For additional background information on Rab proteins.
Specifications
| RAB37 | |
| Polyclonal | |
| Unconjugated | |
| RAB37 | |
| B230331O03Rik; B230354I04Rik; GTPase Rab37; LOC100364984; Rab37; RAB37, member of RAS oncogene family; RAB37, member RAS oncogene family; ras-related protein Rab-37; rCG35079-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100364984, 326624 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96AX2 | |
| RAB37 | |
| A synthesized peptide derived from human RAB37(Accession Q96AX2), corresponding to amino acid residues F169-C219. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction