missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB38 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99623V
This item is not returnable.
View return policy
Description
RAB38 Polyclonal antibody specifically detects RAB38 in Human samples. It is validated for Western Blot
Specifications
| RAB38 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| Melanoma antigen NY-MEL-1, NY-MEL-1, RAB38, member RAS oncogene family, Rab-related GTP-binding protein, ras-related protein Rab-38, rrGTPbp | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RAB38. | |
| 0.1 mL | |
| Signal Transduction | |
| 23682 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction