missing translation for 'onlineSavingsMsg'
Learn More

RAB38 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 216.00 - € 307.65

Specifications

Antigen RAB38
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18604206
View Documents
Novus Biologicals
NBP2-99623-100ul
100 μg
ONLINE EXCLUSIVE PRICE
€ 326.00
€ 307.65 / 100µL
Save € 18.35
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18675555
View Documents
Novus Biologicals
NBP2-99623-50ul
50 μg
€ 216.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RAB38 Polyclonal antibody specifically detects RAB38 in Human samples. It is validated for Western Blot
Specifications

Specifications

RAB38
Western Blot
Unconjugated
Rabbit
Signal Transduction
PBS (pH 7.0)
23682
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
Human
Melanoma antigen NY-MEL-1, NY-MEL-1, RAB38, member RAS oncogene family, Rab-related GTP-binding protein, ras-related protein Rab-38, rrGTPbp
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RAB38.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.