missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106718
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAB38.
May be involved in melanosomal transport and docking. Involved in the proper sorting of TYRP1.
Specifications
| RAB38 | |
| Polyclonal | |
| Unconjugated | |
| Rab38 | |
| 2310011F14Rik; AU043391; chocolate; cht; melanoma antigen NY-MEL-1; NY-MEL-1; R; RAB38; Rab38, member of RAS oncogene family; RAB38, member RAS oncogene family; rab38a; RAB38a, member RAS oncogene family; Rab-related GTP-binding protein; ras-related protein Rab-38; red eyed dilution; rrGTPbp; Ruby; ruby or red eyed dilution; small GTP binding protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 23682, 252916, 72433 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P57729, Q8QZZ8 | |
| Rab38 | |
| A synthesized peptide derived from human RAB38(Accession P57729), corresponding to amino acid residues Y79-C129. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction