missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB39 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93927-0.1ml
This item is not returnable.
View return policy
Description
RAB39 Polyclonal antibody specifically detects RAB39 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RAB39 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| rab-39, RAB39, member RAS oncogene family, RAB39A, rab-related GTP-binding protein, ras-related protein Rab-39A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-170 of human RAB39A (NP_059986.1). SFEHVKDWLEEAKMYVQPFRIVFLLVGHKCDLASQRQVTREEAEKLSADCGMKYIETSAKDATNVEESFTI | |
| 0.1 mL | |
| Signal Transduction | |
| 54734 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction