missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB39B Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97201PCP
This item is not returnable.
View return policy
Description
RAB39B Polyclonal antibody specifically detects RAB39B in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RAB39B | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| MRX72, RAB39B, member RAS oncogene family, ras-related protein Rab-39B | |
| Produced in rabbits immunized with E. coli-derived Human RAB39B fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 116442 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction