missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3A Monoclonal Antibody (OTI6D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809486S
This item is not returnable.
View return policy
Description
RAB3A Monoclonal antibody specifically detects RAB3A in Human samples. It is validated for Western BlotSpecifications
| RAB3A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQE9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9275 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6D10 | |
| Unconjugated | |
| BCL7B | |
| C18A3.6; RAB3; Rab3a; RAB3A, member RAS oncogene family; RAS-associated protein RAB3A; ras-related protein Rab-3A; ras-related protein Rab-3A-like protein; Ras-related small GTP binding protein 3A | |
| Full length human recombinant protein of human BCL7B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction