missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3A Monoclonal Antibody (OTI6D10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809486
This item is not returnable.
View return policy
Description
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.Specifications
| RAB3A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P20336 | |
| RAB3A | |
| Full length human recombit protein of human RAB3A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6D10 | |
| Unconjugated | |
| RAB3A | |
| RAB3A, member RAS oncogene family | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5864 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction