missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAB3a Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14691
This item is not returnable.
View return policy
Description
This antibody is predicted to react with human, mouse, Xenopus and non-human primate based on 100% sequence homology. In Western blot, this antibody is specific for ~27 kDa Rab 3 protein.
Rab proteins belong to the Ras small GTPase superfamily. The GTP-binding Rab3 subfamily has 4 known isoforms: 3A, 3B, 3C, and 3D. These proteins are expressed in highest concentrations in brain and endocrine tissues and involved in regulated exocytosis of hormones and neurotransmitters. Specifically, Rab 3s are thought to play a key role in presynaptic vesicle trafficking and priming.
Specifications
| RAB3a | |
| Polyclonal | |
| Unconjugated | |
| RAB3A | |
| C18A3.6; RAB3; Rab3a; RAB3A, member RAS oncogene family; RAS-associated protein RAB3A; ras-related protein Rab-3A; ras-related protein Rab-3A-like protein; Ras-related small GTP binding protein 3A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19339, 25531, 5864 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| 0.01M HEPES with 50% glycerol, 0.15M NaCl, 100 μg/mL BSA and no preservative, pH 7.5 | |
| P20336, P63011, P63012 | |
| RAB3A | |
| Synthetic peptide corresponding to the N-terminal region of rat RAB3A conjugated to KLH. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction