missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Proteoglycans Septum Cartilage Native Protein

Product Code. 11559071
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11559071 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11559071 Supplier Invitrogen™ Supplier No. RP77523

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Native Protein

Proteoglycans Septum Cartilage (human) Native Protein, RP-77523, from Invitrogen.

RP-77523 is native Proteoglycans Septum Cartilage protein purified from human septum cartilage using Guanidine-HCl extraction, pepsin digestion, ethanol precipitation, potassium-acetate extraction and chromatography. The mixture contains large proteoglycans consisting of core proteins with attached glycosaminoglycans. Major components of the mixture are aggrecan and versican, which are hyaluronic acid-binding proteoglycans. Both contain hyaluronic acid binding regions, Ig-like domains, EGF like domains and lectin-like domains. As macromolecular polyanions they expand and hydrate the ECM in cartilage, intervertebral discs, dermis, blood vessel walls and brain tissue. Aggrecan and versican are degraded by proteases of the ADAMTS family. Aggrecan is cleaved by ADAMTS 1, 4, 5, 8, 9 and 15. Versican is hydrolyzed by ADAMTS 1, 4 and 9. RP-77523 may be used as a control with corresponding anti-Proteoglycans Septum Cartilage antibodies.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control
Formulation protein with no preservative
Name Human Proteoglycans Septum Cartilage
Quantity 1 mg
Source Human
Storage Requirements 4°C or -80°C if preferred
Regulatory Status RUO
Common Name Human Proteoglycans Septum Cartilage
Specific Reactivity Human
Conjugate Unconjugated
Cross Reactivity Hu
Species Human
Recombinant Native
Form Lyophilized
Protein Subtype Native
Purity or Quality Grade purified
Show More Show Less

in vitro research only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.