missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab3D Antibody (5E9), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14764G
This item is not returnable.
View return policy
Description
Rab3D Monoclonal antibody specifically detects Rab3D in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Rab3D | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 5000000000 | |
| Western Blot | |
| D2-2, GOVras-related protein Rab-3D, RAB16glioblastoma overexpressed, RAB3D, member RAS oncogene family, RAD3DRab3D upregulated with myeloid differentiation | |
| Full length recombinant Rab3D protein (Accession #NP_004274) | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Signal Transduction | |
| 9545 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction