missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB3IL1 Antibody (OTI1B5), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73774APCCY7
This item is not returnable.
View return policy
Description
RAB3IL1 Monoclonal specifically detects RAB3IL1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| RAB3IL1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| RAB3IL1 | |
| Full length recombinant protein of human RAB3IL1 (NP_037533) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Flow Cytometry | |
| OTI1B5 | |
| Flow Cytometry | |
| GRAB, guanine nucleotide exchange factor for Rab3A, guanine nucleotide exchange factor for Rab-3A, RAB3A interacting protein (rabin3)-like 1, Rab-3A-interacting-like protein 1, Rab3A-interacting-like protein 1, Rabin3-like 1 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 5866.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction